Merck
  • Comparison of therapeutic lipid target achievements among high-risk patients in Oman.
Comparison of therapeutic lipid target achievements among high-risk patients in Oman.
Angiology (2013-04-09)
Khalid Al-Waili, Ibrahim Al-Zakwani, Tamima Al-Dughaishi, Yajnavalka Baneerje, Hilal Al-Sabti, Khamis Al-Hashmi, Hatem Farhan, Khadija Al Habsi, Ali T Al-Hinai, Khalid Al-Rasadi
ABSTRACT
We compared therapeutic lipid target achievements among patients with diabetes or coronary heart disease (CHD) in Oman. A retrospective chart review of 94 patients was conducted at an outpatient clinic in Sultan Qaboos University Hospital, Muscat, Oman. The variables included low-density lipoprotein cholesterol (LDL-C), high-density lipoprotein cholesterol (HDL-C), and apolipoprotein B (apo B). The overall mean age of the cohort was 59 ± 12 years, 54% were male, 66% were diabetic, 48% hypertensive, 45% had CHD, 94% were on simvastatin, 4% were on fenofibrate, and 2% were on both simvastatin and fenofibrate. Lipid goal attainments of calculated LDL-C (<2.6 mmol/L), apo B (<0.9 g/L), and non-HDL-C (<3.36 mmol/L) were reached in 52%, 39%, and 53% of the patients, respectively. A significant proportion of high-risk patients treated with lipid-lowering agents reach LDL-C but not the apo B treatment targets, suggesting that the use of apo B target values should also be considered.
Product Description
Simvastatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Supelco
Simvastatin, analytical standard
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Simvastatin, United States Pharmacopeia (USP) Reference Standard
USP
Fenofibrate, United States Pharmacopeia (USP) Reference Standard
Fenofibrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fenofibrate, ≥99%, powder
Supelco
Fenofibrate, Pharmaceutical Secondary Standard; Certified Reference Material