Merck
  • Development of an orange juice in-house reference material and its application to guarantee the quality of vitamin C determination in fruits, juices and fruit pulps.
Development of an orange juice in-house reference material and its application to guarantee the quality of vitamin C determination in fruits, juices and fruit pulps.
Food chemistry (2014-02-13)
A Valente, A Sanches-Silva, T G Albuquerque, H S Costa
ABSTRACT
Reference materials are useful for the quality control of analytical procedures and to evaluate the performance of laboratories. There are few and expensive certified reference materials commercially available for vitamin C or ascorbic acid analysis in food matrices. In this study, the preparation and the suitability assessment of an orange juice in-house reference material (RM) for vitamin C analysis in fruits, juices and in fruit pulps is described. This RM was used for the development and full validation of an HPLC method. The results showed excellent linearity (r(2)=0.9995), good accuracy (96.6-97.3%) and precision, as relative standard deviation, ranged from 0.70% to 3.67%. The in-house RM was homogenous and stable at storage conditions (-80°C) during 12 months. According to our results, this in-house RM is an excellent tool to use in quality control and method verification purposes for vitamin C analysis of fruits, juices and fruit pulps matrices. Furthermore, a stabilization solution with perchloric and metaphosphoric acids was developed which prevents degradation of ascorbic acid for a period of 12 months at -80°C.
Product Description
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard