Merck
  • Calculated and experimental chromatograms for distorted gradients and non-linear solvation strength retention models.
Calculated and experimental chromatograms for distorted gradients and non-linear solvation strength retention models.
Journal of chromatography. A (2014-07-08)
Fabrice Gritti, Georges Guiochon
ABSTRACT
Computer calculations of gradient chromatograms were performed by taking into account the adsorption behavior of the strong eluent in RPLC and the true Henry constant of the analytes. This improves the accuracy of classical gradient calculations, which all assume no affinity of the eluent modifier for the stationary phase and that the linear solvation strength model (LSSM) applies. The excess adsorption isotherm of acetonitrile with respect to water was measured by the minor disturbance method onto a Symmetry-C₁₈ RPLC adsorbent. The variations of the Henry constants of a nine compound mixture with the volume fraction of acetonitrile in the aqueous mobile phase were measured. The equilibrium dispersive model of chromatography combined with orthogonal collocation on finite elements was used to calculate chromatograms of the sample mixture for four gradient times decreasing from 25 to 1 min. The results predict a loss of resolution for the less retained analytes when the gradient times becomes smaller than 4 min. They also predict that this behavior can be eliminated when applying a quadratic gradient profile rather than a classical linear gradient. The predictions were validated by the agreement between the calculated and experimental chromatograms.
Product Description
Supelco
Water
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, suitable for HPLC
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
3,5-Dihydroxybenzyl alcohol, 99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)