Merck
  • Removal of phenolic compounds from aqueous phase by adsorption onto polymer supported iron nanoparticles.
Removal of phenolic compounds from aqueous phase by adsorption onto polymer supported iron nanoparticles.
Bulletin of environmental contamination and toxicology (2014-09-27)
Bhupendra K Sen, Dhananjay K Deshmukh, Manas K Deb, Devsharan Verma, Jolly Pal
ABSTRACT
The removal of phenolic compounds, i.e., o-cresol, m-cresol, and p-cresol from aqueous solution have been evaluated employing activated carbon (AC) coated with polymer supported iron nanoparticles (FeNPs). The synthesized FeNPs were characterized by scanning electron microscope and X-ray diffraction analysis. High correlation coefficient values indicated that the adsorption of phenolic compounds onto AC coated with polyvinylpyrrolidon (PVP) supported FeNPs obey Freundlich and Langmuir adsorption isotherms. Higher Freundlich and Langmuir constant values for AC coated with PVP supported FeNPs indicated its greater efficiency than AC. The adsorption data are well represented by both the Freundlich and Langmuir isotherms, indicating favourable adsorption of cresols by the adsorbents. Cresols were effectively removed (90 %) by adsorption process from aqueous solution using AC coated with FeNPs. The percentage removal of above phenolic compounds was studied under varying experimental conditions such as pH, temperature, adsorbent dosage, and contact time. The adsorption of phenolic compounds is quite sensitive to pH of the suspension and optimum uptake value was found at pH 7.0. Temperature also has a favorable effect on adsorption when varied from 20 to 50°C. On the contrary, beyond 30°C, a decrease in the adsorption was noticed.
Product Description
Supelco
Poly(vinylpolypyrrolidone), ~110 μm particle size
Supelco
Water, suitable for ion chromatography
Supelco
Water, for TOC analysis
Sigma-Aldrich
Kollidon® 25
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Povidone, meets USP testing specifications
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, Deionized
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, foil, not light tested, 100x100mm, thickness 0.006mm, 99.85%
Iron, foil, not light tested, 150x150mm, thickness 0.008mm, 99.85%
USP
Crospovidone, United States Pharmacopeia (USP) Reference Standard
Crospovidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Water, ACS reagent
Iron, foil, not light tested, 50x50mm, thickness 0.001mm, 99.85%
Sigma-Aldrich
Water, suitable for HPLC