Merck
  • In vivo tracking and immunological properties of pulsed porcine monocyte-derived dendritic cells.
In vivo tracking and immunological properties of pulsed porcine monocyte-derived dendritic cells.
Molecular immunology (2014-10-06)
Elisa Crisci, Lorenzo Fraile, Rosa Novellas, Yvonne Espada, Raquel Cabezón, Jorge Martínez, Lorena Cordoba, Juan Bárcena, Daniel Benitez-Ribas, María Montoya
ABSTRACT
Cellular therapies using immune cells and in particular dendritic cells (DCs) are being increasingly applied in clinical trials and vaccines. Their success partially depends on accurate delivery of cells to target organs or migration to lymph nodes. Delivery and subsequent migration of cells to regional lymph nodes is essential for effective stimulation of the immune system. Thus, the design of an optimal DC therapy would be improved by optimizing technologies for monitoring DC trafficking. Magnetic resonance imaging (MRI) represents a powerful tool for non-invasive imaging of DC migration in vivo. Domestic pigs share similarities with humans and represent an excellent animal model for immunological studies. The aim of this study was to investigate the possibility using pigs as models for DC tracking in vivo. Porcine monocyte derived DC (MoDC) culture with superparamagnetic iron oxide (SPIO) particles was standardized on the basis of SPIO concentration and culture viability. Phenotype, cytokine production and mixed lymphocyte reaction assay confirmed that porcine SPIO-MoDC culture were similar to mock MoDCs and fully functional in vivo. Alike, similar patterns were obtained in human MoDCs. After subcutaneous inoculation in pigs, porcine SPIO-MoDC migration to regional lymph nodes was detected by MRI and confirmed by Perls staining of draining lymph nodes. Moreover, after one dose of virus-like particles-pulsed MoDCs specific local and systemic responses were confirmed using ELISPOT IFN-γ in pigs. In summary, the results in this work showed that after one single subcutaneous dose of pulsed MoDCs, pigs were able to elicit specific local and systemic immune responses. Additionally, the dynamic imaging of MRI-based DC tracking was shown using SPIO particles. This proof-of-principle study shows the potential of using pigs as a suitable animal model to test DC trafficking with the aim of improving cellular therapies.
Product Description
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
SAFC
L-Glutamine
Dextran 70 for calibration, Dextran 70 European Pharmacopoeia (EP) Reference Standard
Dextran 10 for calibration, Dextran 10 European Pharmacopoeia (EP) Reference Standard
Dextran 40 for calibration, Dextran 40 European Pharmacopoeia (EP) Reference Standard
Dextran 250 for calibration, Dextran 250 European Pharmacopoeia (EP) Reference Standard
Dextran 40 for performance test, Dextran 40 European Pharmacopoeia (EP) Reference Standard
Dextran 60/70 for performance test, Dextran 60/70 European Pharmacopoeia (EP) Reference Standard
Dextran EP Reference Standard, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 25,000
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
Dextran, analytical standard, suitable for gel permeation chromatography (GPC), Set Mp 1,000-400,000
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture