Merck
  • The lipoprotein receptor LRP1 modulates sphingosine-1-phosphate signaling and is essential for vascular development.
The lipoprotein receptor LRP1 modulates sphingosine-1-phosphate signaling and is essential for vascular development.
Development (Cambridge, England) (2014-11-08)
Chikako Nakajima, Philipp Haffner, Sebastian M Goerke, Kai Zurhove, Giselind Adelmann, Michael Frotscher, Joachim Herz, Hans H Bock, Petra May
ABSTRACT
Low density lipoprotein receptor-related protein 1 (LRP1) is indispensable for embryonic development. Comparing different genetically engineered mouse models, we found that expression of Lrp1 is essential in the embryo proper. Loss of LRP1 leads to lethal vascular defects with lack of proper investment with mural cells of both large and small vessels. We further demonstrate that LRP1 modulates Gi-dependent sphingosine-1-phosphate (S1P) signaling and integrates S1P and PDGF-BB signaling pathways, which are both crucial for mural cell recruitment, via its intracellular domain. Loss of LRP1 leads to a lack of S1P-dependent inhibition of RAC1 and loss of constraint of PDGF-BB-induced cell migration. Our studies thus identify LRP1 as a novel player in angiogenesis and in the recruitment and maintenance of mural cells. Moreover, they reveal an unexpected link between lipoprotein receptor and sphingolipid signaling that, in addition to angiogenesis during embryonic development, is of potential importance for other targets of these pathways, such as tumor angiogenesis and inflammatory processes.
Product Description
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
SAFC
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
D-Sphingosine, synthetic
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-LRP, Light Chain Mouse mAb (5A6), liquid, clone 5A6, Calbiochem®
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
(±)-Propylene oxide, ReagentPlus®, ≥99%
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
(±)-Propylene oxide, analytical standard
Sigma-Aldrich
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Sphingosine 1-phosphate, ≥95%, powder
Sigma-Aldrich
Sphingosine 1-phosphate, ≥98.0% (TLC)