Merck
  • Extrolites of Wallemia sebi, a very common fungus in the built environment.
Extrolites of Wallemia sebi, a very common fungus in the built environment.
Indoor air (2014-01-30)
T C Desroches, D R McMullin, J D Miller
ABSTRACT
Wallemia sebi has been primarily known as a spoilage fungus of dried, salted fish and other foods that are salty or sweet. However, this fungus is also very common in house dust. The health effects of chronic exposure to mold and dampness are known to be associated with both allergens and various inflammatory compounds, including the secondary metabolites of building associated fungi and their allergens. IgE sensitization to W. sebi has been long reported from housing and occupational exposures. However, its allergens have not been described previously. Strains from food have been reported to produce a number of compounds with modest toxicity. Strains from the built environment in Canada produced a number of metabolites including the known compound walleminone and a new compound 1-benzylhexahydroimidazo [1,5-α] pyridine-3,5-dione which we call wallimidione. Based on an in silico analysis, wallimidione is likely the most toxic of the metabolites reported to date from W. sebi. We found that the primary human antigen of W. sebi is a 47 kDa excreted cellulase present in high concentrations in W. sebi arthrospores. This species is a basidiomycete and, unsurprisingly, the antigen was not found in extracts of other fungi common in the built environment, all ascomycetes.
Product Description
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with fungal and yeast extracts, DMSO solution
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid solution, suitable for HPLC