Merck
  • A phosphorylation-sensitive tyrosine-tailored magnetic particle for electrochemically probing free organophosphates in blood.
A phosphorylation-sensitive tyrosine-tailored magnetic particle for electrochemically probing free organophosphates in blood.
The Analyst (2014-09-02)
Yanmei Si, Ning Zhang, Zongzhao Sun, Shuai Li, Liyang Zhao, Rui Li, Hua Wang
ABSTRACT
A simple, rapid, sensitive, selective, and field-deployable detection protocol has been initially proposed for the early warning and diagnosis of exposure to organophosphates (OPs) by electrochemically monitoring the direct biomarkers of free OPs in blood. Phosphorylation-sensitive tyrosine (Tyr), which was tested with unique electroactivity, was bound onto Fe3O4 particles mediated by the mussel-inspired dopamine to form Fe3O4@Tyr particles with well-defined shape and well-retained Tyr electroactivities, as characterized separately by electron microscopy and electrochemical measurements. A "lab-on-a-particle"-based detection procedure combined with a magnetic electrode was thus developed by employing Fe3O4@Tyr particles as capturing probes for detecting free OPs in blood, dimethyl-dichloro-vinyl phosphate (DDVP) as an example. A significant difference in electrochemical responses could be obtained for Fe3O4@Tyr particles before and after DDVP exposure, based on the phosphorylation-induced inhibition of electroactivities of loaded Tyr. Investigation results indicate that highly specific and sensitive phosphorylation for the inhibition of Tyr electroactivities by sensitive electrochemical outputs could endow the OP detection with high selectivity and sensitivity (i.e., down to about 0.16 nM DDVP in blood). Moreover, strong and stable Tyr-OP bindings especially irreversible electrochemical oxidization of the Tyr probe could facilitate the OP evaluation with high reproducibility and stability over time. In particular, the simple "lab-on-a-particle"-based detection procedure equipped with a portable electrochemical transducer can be tailored for the field-deployable or on-site monitoring of the exposure to various nerve agents and pesticides.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Ascorbic acid, analytical standard
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
SAFC
Glycine
SAFC
L-Threonine
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)