Merck
  • Molecular characterization of p.Asp77Gly and the novel p.Ala163Val and p.Ala163Glu mutations causing protein C deficiency.
Molecular characterization of p.Asp77Gly and the novel p.Ala163Val and p.Ala163Glu mutations causing protein C deficiency.
Thrombosis research (2015-01-27)
Kitti B Kovács, István Pataki, Helga Bárdos, Attila Fekete, György Pfliegler, Gizella Haramura, Réka Gindele, István Komáromi, György Balla, Róza Ádány, László Muszbek, Zsuzsanna Bereczky
ABSTRACT
Protein C (PC) is a major anticoagulant and numerous distinct mutations in its coding gene result in quantitative or qualitative PC deficiency with high thrombosis risk. Homozygous or compound heterozygous PC deficiency usually leads to life-threatening thrombosis in neonates. The molecular consequences of 3 different missense mutations of two patients have been investigated. The first patient suffered from neonatal purpura fulminans and was a compound heterozygote for p.Asp77Gly and p.Ala163Glu mutations. The second patient had severe deep venous thrombosis in young adulthood and carried the p.Ala163Val mutation. The fate of mutant proteins expressed in HEK cells was monitored by ELISA, by Western blotting, by investigation of polyubiquitination and by functional assays. Their intracellular localization was examined by immunostaining and confocal laser scanning microscopy. Molecular modeling and dynamics simulations were also carried out. The 163Val and 163Glu mutants had undetectable levels in the culture media, showed intracellular co-localization with the 26S proteasome and were polyubiquitinated. The 77Gly mutant was secreted to the media showing similar activity as the wild type. There was no difference among intracellular PC levels of wild type and mutant proteins. The 163Val and 163Glu mutations caused significant changes in the relative positions of the EGF2 domains suggesting misfolding with the consequence of secretion defect. No major structural alteration was observed in case of 77Gly mutant; it might influence the stability of protein complexes in which PC participates and may have an impact on the clearance of PC requiring further research.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Millipore
D-(+)-Mannose, ≥99%, suitable for microbiology
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)