Merck
CREG promotes vasculogenesis by activation of VEGF/PI3K/Akt pathway.
Frontiers in bioscience (Landmark edition) (2014-06-05)
Xiaoxiang Tian, Na Zhang, Chenghui Yan, Jun Nelsen, Shaowei Liu, Jian Kang, Jian Zhang, Chengfei Peng, Jie Tao, Mingyu Sun, Yang Li, Shaohua Li, Hong Wang, Yaling Han
ABSTRACT
Knowledge about factors regulating vasculogenesis remains limited. The cellular repressor of E1A-stimulated gene (CREG) has been reported to be involved in maintaining cellular differentiation and endothelial homeostasis, thus we hypothesize that CREG may be a novel factor regulating vasculogenesis. By using mouse embryonic stem cells (ESC) derived embryoid body (EB) model, we confirmed expression of CREG was significantly up-regulated during EB differentiation. Overexpression of CREG in ESC led to accelerated cystic EB formation, increased endothelial differentiation and vasculogenesis, whereas knockdown of CREG produced opposite phenotypes. Moreover, we found expression of vascular endothelial growth factor (VEGF) was up-regulated and PI3K/Akt pathway was activated in CREG-overexpressing EB. Administration of VEGF neutralizing antibody or PI3K/Akt pharmacological inhibitor LY294002 blocked the vasculogenesis in CREG over-expressing EB, while supplement of VEGF rescued vasculogenesis deficiency in CREG knocked down EB. Further study by Western blot determined that PI3K/Akt was a downstream effector of VEGF. We identify CREG as a novel factor in regulating endothelial differentiation and vasculogenesis via VEGF/PI3K/Akt pathway.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Creg1
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Human VEGF ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human VEGF R2 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human CREG1 (2)
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Mouse VEGF ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat VEGF ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Rat VEGF ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse VEGF ELISA Kit, for serum, plasma, cell culture supernatant
Sigma-Aldrich
Human VEGF R3 ELISA Kit, for serum, plasma, cell culture supernatant, urine
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material