Merck
  • Endothelium-dependent and -independent relaxation of rat aorta induced by extract of Schizophyllum commune.
Endothelium-dependent and -independent relaxation of rat aorta induced by extract of Schizophyllum commune.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2014-08-31)
Haiyun Chen, Sujuan Li, Peng Wang, Saimei Yan, Lin Hu, Xiaoxia Pan, Cui Yang, George Pakheng Leung
ABSTRACT
Schizophyllum commune (SC) is widely consumed by Chinese, especially in southern part of China. The aim of the present study was to assess the extract of SC on vascular tone and the mechanisms involved. Experiments were performed on aorta of 18-week-old male Sprague-Dawley rats. Dried SC was extracted with 50% ethanol, 90% ethanol and deionized water, respectively. The effects of SC on the isometric tension of rat aortic rings were measured. Protein expression for the endothelial nitric oxide synthase (eNOS) was also determined in the primarily cultured rat aortic arterial endothelial cells (RAECs). The results showed that the water extract of SC induced a marked relaxation in aortic rings with or without endothelium. After the pretreatments of N(ω)-nitro-l-arginine methyl ester, indomethacin, RP-cAMP, and methylene blue, the SC-induced relaxation was significantly decreased. In addition, the contraction due to Ca(2+) influx and intracellular Ca(2+) release was also inhibited by SC. Furthermore, expression of the eNOS protein was significantly elevated in RAECs after treatment of SC. In conclusion, the water extract of SC induces an endothelium-dependent and -independent relaxation in rat aorta. The relaxing effect of SC involves the modulation of NO-cGMP-dependent pathways, PGI2-cAMP-depedent pathways, Ca(2+) influx though calcium channels and intracellular Ca(2+) release.
Sigma-Aldrich
3-[(1R)-1-Hydroxy-2-(methylamino)ethyl]phenol, AldrichCPR
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Adenosine, European Pharmacopoeia (EP) Reference Standard
Phenylephrine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenosine