Merck
  • Capillary electromigration techniques as tools for assessing the status of vitamins A, C and E in patients with cystic fibrosis.
Capillary electromigration techniques as tools for assessing the status of vitamins A, C and E in patients with cystic fibrosis.
Journal of pharmaceutical and biomedical analysis (2014-09-23)
Ilona Olędzka, Katarzyna Kaźmierska, Alina Plenis, Barbara Kamińska, Tomasz Bączek
ABSTRACT
The purpose of this work is the evaluation of the nutritional status of patients with cystic fibrosis (CF), based on the level of vitamin C in urine and vitamins A and E in serum, using the fast, selective and fully automated micellar electrokinetic capillary chromatographic (MEKC) and microemulsion electrokinetic capillary chromatographic (MEEKC) methods. The optimization of parameters affecting the electrophoretic separation provided adequate separation of the analytes of interest in the short time of 8 min (MEKC) and 20 min (MEEKC). The developed methods were practical applications to evaluate the levels of vitamins A, C and E in real samples from 28 children suffering from cystic fibrosis and from 10 healthy volunteers. Based on the mean concentration values obtained in the two groups, it can be seen that the levels of each vitamin were lower in patients with CF than in healthy volunteers. In the case of vitamin E, these differences in both groups were statistically significant, while the disproportion of concentrations of vitamins A and C in both the studied groups were not so relevant. On the other hand, a principal component analysis (PCA) confirmed that in some patients with CF the concentration of vitamin A was significantly lower than in the control group. Thus, the future evaluation of the status of fat-soluble vitamins in the longer term for the evaluation of the nutritional status of patients with CF should be continued. The presented CE methods can become useful tools for the evaluation of the nutritional status of patients with CF.
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Thiamine hydrochloride, United States Pharmacopeia (USP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Supelco
1-Butanol, analytical standard
Supelco
Octane, analytical standard
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Thiamine hydrochloride, ≥98%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Thiamine hydrochloride, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Octane, anhydrous, ≥99%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade