Merck
  • Role of SIRT1 in regulation of epithelial-to-mesenchymal transition in oral squamous cell carcinoma metastasis.
Role of SIRT1 in regulation of epithelial-to-mesenchymal transition in oral squamous cell carcinoma metastasis.
Molecular cancer (2014-11-27)
I-Chieh Chen, Wei-Fan Chiang, Hsin-Hsiu Huang, Pei-Fen Chen, Ying-Ying Shen, Hung-Che Chiang
ABSTRACT
The epithelial-to-mesenchymal transition (EMT) process results in a loss of cell-cell adhesion, increased cell mobility, and is crucial for enabling the metastasis of cancer cells. Recently, the enzyme SIRT1 has been implicated in a variety of physiological processes; however, its role in regulating oral cancer metastasis and EMT is not fully elucidated. Here, we propose a mechanism by which the enzyme sirtuin1 (SIRT1) regulates the EMT process in oral cancer by deacetylating Smad4 and repressing the effect of TGF-β signaling on matrix metalloproteinase-7 (MMP7). The roles of SIRT1 in tumor cell migration/invasion and metastasis to the lungs were investigated using the Boyden chamber assay and orthotopic injections, respectively. RNA interference was used to knockdown either SIRT1 or Smad4 expression in oral squamous cell carcinoma (OSCC) cell lines. Immunoblotting, zymographic assays, and co-immunoprecipitation were used to examine the effects of SIRT1 overexpression on MMP7 expression and activity, as well as on SIRT1/ Smad4 interaction. We found that compared with normal human oral keratinocytes (HOKs), SIRT1 was underexpressed in OSCC cells, and also in oral cancer tissues obtained from 14 of 21 OSCC patients compared with expression in their matched normal tissues. Overexpression of SIRT1 inhibited migration of OSCC cells in vitro, as well as their metastasis to the lung in vivo. Furthermore, up-regulation of SIRT1 in metastatic OSCCs significantly inhibited the migration and invasion abilities of OSCC cells, while concomitantly increasing the expression of E-cadherin, and decreasing the expressions of mesenchymal markers. We also identified Smad4, a TGF-β-activated transcription factor, as a direct target protein for SIRT1. Overexpression of SIRT1 in OSCC cells led to decreased levels of acetylated Smad4, and inhibition of TGF-β-induced signaling. By associating and deacetylating Smad4, SIRT1 enzyme can influence MMP7 expression, MMP enzyme activity, and consequently, cell migration, invasion, and tumor metastasis in OSCCs. These findings provide a valuable insight into the potential role of the SIRT1 enzyme in regulating cell migration and invasion in oral squamous cell carcinoma. Our findings suggest the SIRT1/Smad4/MMP7 pathway as a target for oral cancer driven by EMT.
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sirtinol, ≥95% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder