Merck
  • Effect of low frequency (LF) electric fields on gene expression of a bone human cell line.
Effect of low frequency (LF) electric fields on gene expression of a bone human cell line.
Electromagnetic biology and medicine (2013-08-28)
Mariella Caputo, Hylde Zirpoli, Maria Caterina De Rosa, Tania Rescigno, Francesco Chiadini, Antonio Scaglione, Claudia Stellato, Giorgio Giurato, Alessandro Weisz, Mario Felice Tecce, Bruno Bisceglia
ABSTRACT
We evaluated the effects, on cultured human SaOS-2 cells, of exposures to the low frequency (LF) electric signal (60 kHz sinusoidal wave, 24.5 V peak-to-peak voltage, amplitude modulated by a 12.5 Hz square wave, 50% duty cycle) from an apparatus of current clinical use in bone diseases requiring regenerating processes. Cells in flasks were exposed to a capacitively coupled electric field giving electric current density in the sample of 4 µA/cm(2). The whole expressed cellular mRNAs were systematically analyzed by "DNA microchips" technology to identify all individual species quantitatively affected by field exposure. Comparisons were made between RNA samples from exposed and control sham-exposed cells. Results indicated that immediately and 4 h after exposure there were almost no differentially modulated mRNA species. However, samples obtained at 24 h after exposure showed a small number of limitedly differential signals (7 down-regulated and 3 up-regulated with a cut-off value of ±1.5; 38 and 11, respectively, with a cut-off value of ±1.3), which included mostly mRNA encoding transcription factors and DNA binding proteins. Nevertheless, in identical experimental conditions, we previously demonstrated enzymatic changes of alkaline phosphatase occurring immediately after exposure and declining in a few hours. Therefore, since enzymatic changes occur before those observed at gene regulation level, it is conceivable that only earlier effects are directly due the treatment and then these effects are later able to affect gene expression only indirectly.
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland