Merck
  • Adenosine, lidocaine and Mg2+ (ALM) induces a reversible hypotensive state, reduces lung edema and prevents coagulopathy in the rat model of polymicrobial sepsis.
Adenosine, lidocaine and Mg2+ (ALM) induces a reversible hypotensive state, reduces lung edema and prevents coagulopathy in the rat model of polymicrobial sepsis.
The journal of trauma and acute care surgery (2014-08-28)
Maddison J Griffin, Hayley L Letson, Geoffrey P Dobson
ABSTRACT
No drug therapy has demonstrated improved clinical outcomes in the treatment of sepsis. A bolus of adenosine, lidocaine, and magnesium (ALM) has been shown to be cardioprotective and restore coagulopathy in different trauma states. We hypothesized that ALM therapy may improve hemodynamics, protect the lung, and prevent coagulopathy in the rat sepsis model. Nonheparinized, anesthetized Sprague-Dawley rats (350-450 g, n = 32) were randomly assigned to (1) shams (without sepsis), (2) saline controls, and (3) ALM treatment. Sepsis was induced by cecal ligation and puncture. A 0.3-mL bolus was administered intravenously, followed by a 4-hour intravenous infusion (1 mL/kg/h), and hemodynamics (mean arterial pressure [MAP], systolic arterial pressure, diastolic arterial pressure, and heart rate [HR]) and body temperature (BT) were monitored. Coagulation was assessed using prothrombin time and activated partial thromboplastin time (aPTT). Shams displayed progressive falls in MAP, HR, and BT as well as a prolonged aPTT, which were related to surgery, not infection. At 4 hours, controls showed more pronounced falls in MAP (33%), HR (17%), and BT (3.3°C), and MAP continued to fall after the infusion was stopped. In contrast, ALM treatment resulted in a rapid fall in MAP from 111 mm Hg to 73 mm Hg at 30 minutes (p < 0.05 all groups) and was 59 mm Hg at 240 minutes (p < 0.05 shams), which immediately corrected after 4 hours (p < 0.05 controls). HR paralleled MAP changes in ALM rats, and BT was significantly higher than that of the controls but not that of shams. ALM rats had no arrhythmias compared with the controls or shams and had significantly lower lung wet-dry ratios. Prothrombin time in saline controls at 1 hour and 5 hours was prolonged but not in the shams or ALM rats. aPTT at 1 hour in the sham, control, and ALM groups was 158 ± 41 seconds, 161 ± 41 seconds, and 54 ± 23 seconds and at 5 hours was 104 ± 43 seconds, 205 ± 40 seconds, and 33 ± 3 seconds (p < 0.05), respectively. An ALM bolus/infusion induces a stable, hypotensive hemodynamic state with no arrhythmias, significantly less pulmonary edema, and a higher BT and prevents coagulopathy compared with the controls.
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine, ≥99%
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M