Merck
  • L-selenomethionine reduces platinum(IV) anticancer model compounds at strikingly faster rates than L-methionine.
L-selenomethionine reduces platinum(IV) anticancer model compounds at strikingly faster rates than L-methionine.
Dalton transactions (Cambridge, England : 2003) (2014-07-31)
Shuying Huo, Jingran Dong, Shigang Shen, Yanli Ren, Changying Song, Jianzhong Xu, Tiesheng Shi
ABSTRACT
L-Selenomethionine (SeMet), the predominant form of selenium acquired from the diet by humans, has been used as a supplement, and exhibit some important functions like cancer prevention and antioxidative defense. Its interactions with Pt(II) anticancer drugs have been characterized, but its redox reactions with platinum(IV) anticancer prodrugs have not been exploited. In this work, the oxidation of SeMet by Pt(IV) anticancer model compounds trans-[PtX2(CN)4](2-) (X = Cl, Br) was characterized. A stopped-flow spectrometer was used to record the rapid scan spectra and to follow the reaction kinetics over a wide pH range. An overall second-order rate law was derived: -d[Pt(IV)]/dt = k'[Pt(IV)][SeMet], where k' pertains to the observed second-order rate constants. The k'-pH profiles showed that k' increased only about 6 times even though the solution pH was varied from 0.25 to 10.5. The redox stoichiometry was determined as Δ[Pt(IV)]/Δ[SeMet] = 1 : (1.07 ± 0.07), suggesting that SeMet was oxidized to selenomethionine selenoxide. The selenoxide together with its hydrated form was identified explicitly by high resolution mass spectral analysis. A reaction mechanism was proposed which encompassed three parallel rate-determining steps relying on the protolytic species of SeMet. Rate constants of the rate-determining steps were obtained from the simulations of the k'-pH profiles. Activation parameters were determined for the reactions of the zwitterionic form of SeMet with the Pt(IV) complexes. A bridged electron transfer process is delineated in the rate-determining steps and several lines of evidence support the bridged electron transfer mode. Strikingly, reduction of [PtX2(CN)4](2-) by SeMet is 3.7 × 10(3)-5.7 × 10(4) times faster than that by L-methionine. Some potential biological consequences resulting from the strikingly fast reduction are discussed.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture