Merck
  • A dihydroselenoquinazoline inhibits S6 ribosomal protein signalling, induces apoptosis and inhibits autophagy in MCF-7 cells.
A dihydroselenoquinazoline inhibits S6 ribosomal protein signalling, induces apoptosis and inhibits autophagy in MCF-7 cells.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2014-07-11)
Esther Moreno, Dahlia Doughty-Shenton, Daniel Plano, María Font, Ignacio Encío, Juan Antonio Palop, Carmen Sanmartín
ABSTRACT
The PI3K/Akt/mTOR/S6 ribosomal protein signalling pathway is a key potential target in breast cancer therapy, playing a central role in proliferation and cell survival. In this study, we found that the seleno-compound 2,4-dihydroselenoquinazoline (3a) generally inhibited this signalling axis in MCF-7 breast cancer cells and caused downregulation of S6 ribosomal protein phosphorylation in a dose- and time-dependent manner. Furthermore, 3a caused a dose- and time-dependent decrease in MCF-7 cell viability as well as cell cycle arrest in G2/M. Interestingly 3a also induced apoptosis, as evidenced by cleavage of PARP and caspase-7, and inhibited autophagy, as demonstrated by accumulation of LC3-II and p62/SQSTM1. Given that induction of autophagy has been previously described as a mechanism by which some breast cancer cells counteract proapoptotic signalling and develop resistance to anti-hormone therapy, this suggests that this derivative, which both triggers apoptosis and inhibits autophagy, may be beneficial in preventing and overcoming resistance in breast cancer cells. The data also show the complexity of this signalling axis which is far from being understood.
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium fluoride, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%