Merck
  • Reduced melanocortin production causes sexual dysfunction in male mice with POMC neuronal insulin and leptin insensitivity.
Reduced melanocortin production causes sexual dysfunction in male mice with POMC neuronal insulin and leptin insensitivity.
Endocrinology (2015-01-16)
Latrice D Faulkner, Abigail R Dowling, Ronald C Stuart, Eduardo A Nillni, Jennifer W Hill
ABSTRACT
Proopiomelanocortin (POMC)-derived peptides like α-melanocyte-stimulating hormone (MSH) substantially improve hepatic insulin sensitivity and regulate energy expenditure. Melanocortinergic agents are also powerful inducers of sexual arousal that are being investigated for a possible therapeutic role in erectile dysfunction. It is currently unclear whether reduced melanocortin (MC) activity may contribute to the sexual dysfunction accompanying obesity and type 2 diabetes. Male rodents with leptin and insulin resistance targeted to POMC neurons (leptin receptor [LepR]/insulin receptor [IR]POMC mice) exhibit obesity, hyperinsulinemia, hyperglycemia, and systemic insulin resistance. In this study, we demonstrate that LepR/IRPOMC males are also subfertile due to dramatic alterations in sexual behavior. Remarkably, these reproductive changes are accompanied by decreased α-MSH production not present when a single receptor type is deleted. Unexpectedly, behavioral sensitivity to α-MSH and MC receptor expression are also reduced in LepR/IRPOMC males, a potential adaptation of the MC system to altered α-MSH production. Together, these results suggest that concurrent insulin and leptin resistance in POMC neurons in individuals with obesity or type 2 diabetes can reduce endogenous α-MSH levels and impair sexual function.
Product Description
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Luteinizing hormone releasing hormone, ≥97% (HPLC)
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Supelco
N,O-Bis(trimethylsilyl)acetamide, derivatization grade (GC derivatization), LiChropur, ≥98.5% (GC)
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Testosterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Luteinizing hormone releasing hormone salmon, ≥97% (HPLC)
Sigma-Aldrich
α-Melanocyte stimulating hormone, ≥97% (HPLC)
Sigma-Aldrich
Luteinizing hormone releasing hormone human acetate salt, ≥98% (HPLC), powder
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG