Merck
  • Metabotropic glutamate receptor 5 knockout reduces cognitive impairment and pathogenesis in a mouse model of Alzheimer's disease.
Metabotropic glutamate receptor 5 knockout reduces cognitive impairment and pathogenesis in a mouse model of Alzheimer's disease.
Molecular brain (2014-06-03)
Alison Hamilton, Jessica L Esseltine, Rebecca A DeVries, Sean P Cregan, Stephen S G Ferguson
ABSTRACT
Alzheimer's disease (AD) pathology occurs in part as the result of excessive production of β-amyloid (Aβ). Metabotropic glutamate receptor 5 (mGluR5) is now considered a receptor for Aβ and consequently contributes to pathogenic Aβ signaling in AD. Genetic deletion of mGluR5 rescues the spatial learning deficits observed in APPswe/PS1ΔE9 AD mice. Moreover, both Aβ oligomer formation and Aβ plaque number are reduced in APPswe/PS1ΔE9 mice lacking mGluR5 expression. In addition to the observed increase in Aβ oligomers and plaques in APPswe/PS1ΔE9 mice, we found that both mTOR phosphorylation and fragile X mental retardation protein (FMRP) expression were increased in these mice. Genetic deletion of mGluR5 reduced Aβ oligomers, plaques, mTOR phosphorylation and FMRP expression in APPswe/PS1ΔE9 mice. Thus, we propose that Aβ activation of mGluR5 appears to initiate a positive feedback loop resulting in increased Aβ formation and AD pathology in APPswe/PS1ΔE9 mice via mechanism that is regulated by FMRP.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Inositol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material