Merck
  • Increased aortic stiffness elevates pulse and mean pressure and compromises endothelial function in Wistar rats.
Increased aortic stiffness elevates pulse and mean pressure and compromises endothelial function in Wistar rats.
American journal of physiology. Heart and circulatory physiology (2014-07-20)
Xiaomei Guo, Xiao Lu, Junrong Yang, Ghassan S Kassab
ABSTRACT
An increase in pulse pressure (PP) is highly associated with hypertension. The goal of this study was to determine the effect of increased aortic stiffness on PP and endothelial dysfunction as precursors to hypertension. A rat model of suddenly increased aortic stiffness by use of a nonconstrictive restraint (glue coating) on aortic surface was created to investigate the change of PP and mean arterial pressure (MAP). Group I (n = 16) underwent aorta restraint for 4 wk. Group II (n = 12) underwent aortic restraint for 4 wk, followed by restraint removal to evaluate extent of reversibility for additional 4 wk. The aortic and peripheral endothelial function was assessed by ACh-stimulated endothelium-dependent vasodilation. The level of nitrate/nitrite (NOx), endothelin-1 (ET-1), and prostacyclin (PGI2) were measured in the serum and artery tissue. We found that aortic stiffening causes a significant increase in PP and MAP (P < 0.05). The endothelial function was markedly blunted (P < 0.05) in both aorta and small peripheral artery. After removal of the restraint, the impaired endothelium function persisted in the aorta likely due to sustained deterioration of aortic wall, but was partially restored in peripheral artery. The endothelial dysfunction was correlated with a decrease in NOx and PGI2 (P < 0.05) and an increase in ET-1 (P < 0.05). Our results show that aortic stiffening results in widening of PP, which affected endothelium function through changes in synthesis of NOx, ET-1, and PGI2. These findings suggest that increased aortic stiffness may be a cause of increased PP and a precursor to hypertension.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material