Merck
  • Separate cellular localizations of human T-lymphotropic virus 1 (HTLV-1) Env and glucose transporter type 1 (GLUT1) are required for HTLV-1 Env-mediated fusion and infection.
Separate cellular localizations of human T-lymphotropic virus 1 (HTLV-1) Env and glucose transporter type 1 (GLUT1) are required for HTLV-1 Env-mediated fusion and infection.
Journal of virology (2014-10-24)
Yosuke Maeda, Hiromi Terasawa, Yuetsu Tanaka, Chisho Mitsuura, Kaori Nakashima, Keisuke Yusa, Shinji Harada
ABSTRACT
Interaction of the envelope glycoprotein (Env) of human T-lymphotropic virus 1 (HTLV-1) with the glucose transporter type 1 (GLUT1) expressed in target cells is essential for viral entry. This study found that the expression level of GLUT1 in virus-producing 293T cells was inversely correlated with HTLV-1 Env-mediated fusion activity and infectivity. Chimeric studies between GLUT1 and GLUT3 indicated that the extracellular loop 6 (ECL6) of GLUT1 is important for the inhibition of cell-cell fusion mediated by Env. When GLUT1 was translocated into the plasma membrane from intracellular storage sites by bafilomycin A1 (BFLA1) treatment in 293T cells, HTLV-1 Env-mediated cell fusion and infection also were inhibited without the overexpression of GLUT1, indicating that the localization of GLUT1 in intracellular compartments rather than in the plasma membrane is crucial for the fusion activity of HTLV-1 Env. Immunoprecipitation and laser scanning confocal microscopic analyses indicated that under normal conditions, HTLV-1 Env and GLUT1 do not colocalize or interact. BFLA1 treatment induced this colocalization and interaction, indicating that GLUT1 normally accumulates in intracellular compartments separate from that of Env. Western blot analyses of FLAG-tagged HTLV-1 Env in virus-producing cells and the incorporation of HTLV-1 Env in virus-like particles (VLPs) indicate that the processing of Env is inhibited by either overexpression of GLUT1 or BFLA1 treatment in virus-producing 293T cells. This inhibition probably is due to the interaction of the Env with GLUT1 in intracellular compartments. Taken together, separate intracellular localizations of GLUT1 and HTLV-1 Env are required for the fusion activity and infectivity of HTLV-1 Env. The deltaretrovirus HTLV-1 is a causative agent of adult T-cell leukemia (ATL) and HTLV-1-associated myelopathy/tropical spastic paraparesis (HAM/TSP). Although HTLV-1 is a complex retrovirus that has accessory genes, no HTLV-1 gene product has yet been shown to regulate its receptor GLUT1 in virus-producing cells. In this study, we found that a large amount of GLUT1 or translocation of GLUT1 to the plasma membrane from intracellular compartments in virus-producing cells enhances the colocalization and interaction of GLUT1 with HTLV-1 Env, leading to the inhibition of cell fusion activity and infectivity. The results of our study suggest that GLUT1 normally accumulates in separate intracellular compartments from Env, which is indeed required for the proper processing of Env.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)