Merck
  • Effect of UV-C radiation on bioactive compounds of pineapple (Ananas comosus L. Merr.) by-products.
Effect of UV-C radiation on bioactive compounds of pineapple (Ananas comosus L. Merr.) by-products.
Journal of the science of food and agriculture (2014-05-24)
Ana Freitas, Margarida Moldão-Martins, Helena S Costa, Tânia G Albuquerque, Ana Valente, Ana Sanches-Silva
ABSTRACT
The industrial processing of pineapple generates a high quantity of by-products. To reduce the environmental impact of these by-products and the inherent cost of their treatment, it is important to characterise and valorise these products, converting them into high added value products. Ultra-violet radiation is one of the main sustainable sanitation techniques for fruits. Since this radiation can induce plant stress which can promote the biosynthesis of bioactive compounds, it is important to evaluate its effect in fruits. The amounts of vitamins (C and E) and carotenoids (α-carotene, β-carotene, β-cryptoxanthin, lutein, lycopene, neoxanthin, violaxanthin and zeaxanthin) in pineapple by-products (core and rind) were analysed before and after treatment with UV radiation. All treated and untreated pineapple by-products contained β-carotene as the main carotenoid (rind, 2537-3225 µg; and core, 960-994 µg 100 g(-1) DW). Pineapple rind also contained lutein (288-297 µg 100 g(-1) DW) and α-carotene (89-126 µg 100 g(-1) DW). The results provide evidence of the potential of pineapple by-products as a source of bioactive compounds with antioxidant activity, which can be used by pharmaceutical, cosmetics and food industries. In addition, UV-C was shown to be a treatment that can add nutritional value to pineapple by-products.
Product Description
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Ammonium dihydrogenphosphate, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Ammonium phosphate monobasic, suitable for plant cell culture
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Ammonium phosphate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis
Supelco
Ammonium phosphate monobasic, analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%
Supelco
Lutein, analytical standard
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Ammonium phosphate monobasic, ReagentPlus®, ≥98.5%
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Ammonium phosphate monobasic, ACS reagent, ≥98%