Merck
  • Cyclosporine and methotrexate-related pharmacogenomic predictors of acute graft-versus-host disease.
Cyclosporine and methotrexate-related pharmacogenomic predictors of acute graft-versus-host disease.
Haematologica (2014-11-27)
Isabelle Laverdière, Chantal Guillemette, Ryad Tamouza, Pascale Loiseau, Regis Peffault de Latour, Marie Robin, Félix Couture, Alain Filion, Marc Lalancette, Alan Tourancheau, Dominique Charron, Gérard Socié, Éric Lévesque
ABSTRACT
Effective immunosuppression is mandatory to prevent graft-versus-host disease and to achieve a successful clinical outcome of hematopoietic stem cell transplantation. Here we tested whether germline single nucleotide polymorphisms in 20 candidate genes related to methotrexate and cyclosporine metabolism and activity influence the incidence of graft-versus-host disease in patients who undergo stem cell transplantation for hematologic disorders. Recipient genetic status of the adenosine triphosphate-binding cassette sub-family C1 and adenosine triphosphate-binding cassette sub-family C2 transporters, 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/ inosine monophosphate cyclohydrolase within the methotrexate pathway, and nuclear factor of activated T cells (cytoplasmic 1) loci exhibit a remarkable influence on severe acute graft-versus-host disease prevalence. Indeed, an increased risk of acute graft-versus-host disease was observed in association with single nucleotide polymorphisms located in 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/inosine monophosphate cyclohydrolase (hazard ratio=3.04; P=0.002), nuclear factor of activated T cells (cytoplasmic 1) (hazard ratio=2.69; P=0.004), adenosine triphosphate-binding cassette sub-family C2 (hazard ratio=3.53; P=0.0018) and adenosine triphosphate-binding cassette sub-family C1 (hazard ratio=3.67; P=0.0005). While donor single nucleotide polymorphisms of dihydrofolate reductase and solute carrier family 19 (member 1) genes are associated with a reduced risk of acute graft-versus-host disease (hazard ratio=0.32-0.41; P=0.0009-0.008), those of nuclear factor of activated T cells (cytoplasmic 2) are found to increase such risk (hazard ratio=3.85; P=0.0004). None of the tested single nucleotide polymorphisms was associated with the occurrence of chronic graft-versus-host disease. In conclusion, by targeting drug-related biologically relevant genes, this work emphasizes the potential role of germline biomarkers in predicting acute graft-versus-host disease. Further investigations are warranted to improve our understanding of these relationships to personalize immunosuppressive therapy and optimize outcomes.
Product Description
Serine, European Pharmacopoeia (EP) Reference Standard
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
(±)-CPP, solid
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Ciclosporin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Methotrexate, European Pharmacopoeia (EP) Reference Standard
SAFC
Methotrexate
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosine, European Pharmacopoeia (EP) Reference Standard