Merck
  • Amplification of human platelet activation by surface pannexin-1 channels.
Amplification of human platelet activation by surface pannexin-1 channels.
Journal of thrombosis and haemostasis : JTH (2014-03-25)
K A Taylor, J R Wright, C Vial, R J Evans, M P Mahaut-Smith
ABSTRACT
Pannexin-1 (Panx1) forms an anion-selective channel with a permeability up to ~1 kDa and represents a non-lytic, non-vesicular ATP release pathway in erythrocytes, leukocytes and neurons. Related connexin gap junction proteins have been reported in platelets; however, the expression and function of the pannexins remain unknown. To determine the expression and function of pannexins in human plate-lets, using molecular, cellular and functional techniques. Panx1 expression in human platelets was det-ermined using qPCR and antibody-based techniques. Contributions of Panx1 to agonist-evoked efflux of cytoplasmic calcein, Ca(2+) influx, ATP release and aggregation were assessed in washed platelets under conditions where the P2X1 receptor response was preserved (0.32 U mL(-1) apyrase). Thrombus formation in whole blood was assessed in vitro using a shear chamber assay. Two structurally unrelated and widely used Panx1 inhibitors, probenecid and carbenoxolone, were used throughout this study, at concentrations that do not affect connexin channels. PANX1, but not PANX2 or PANX3, mRNA was detected in human platelets. Furthermore, Panx1 protein is glycosylated and present on the plasma membrane of platelets, and displays weak physical association with P2X1 receptors. Panx1 inhibition blocked thrombin-evoked efflux of calcein, and reduced Ca(2+) influx, ATP release, platelet aggregation and thrombus formation under arterial shear rates in vitro. The Panx1-dependent contribution was not additive to that of P2X1 receptors. Panx1 is expressed on human platelets and amplifies Ca(2+) influx, ATP release and aggregation through the secondary activation of P2X1 receptors. We propose that Panx1 represents a novel target for the management of arterial thrombosis.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)