Merck
  • Nitric oxide mediates metabolic coupling of omentum-derived adipose stroma to ovarian and endometrial cancer cells.
Nitric oxide mediates metabolic coupling of omentum-derived adipose stroma to ovarian and endometrial cancer cells.
Cancer research (2014-11-27)
Bahar Salimian Rizi, Christine Caneba, Aleksandra Nowicka, Ahmad W Nabiyar, Xinran Liu, Kevin Chen, Ann Klopp, Deepak Nagrath
ABSTRACT
Omental adipose stromal cells (O-ASC) are a multipotent population of mesenchymal stem cells contained in the omentum tissue that promote endometrial and ovarian tumor proliferation, migration, and drug resistance. The mechanistic underpinnings of O-ASCs' role in tumor progression and growth are unclear. Here, we propose a novel nitric oxide (NO)-mediated metabolic coupling between O-ASCs and gynecologic cancer cells in which O-ASCs support NO homeostasis in malignant cells. NO is synthesized endogenously by the conversion of l-arginine into citrulline through nitric oxide synthase (NOS). Through arginine depletion in the media using l-arginase and NOS inhibition in cancer cells using N(G)-nitro-l-arginine methyl ester (l-NAME), we demonstrate that patient-derived O-ASCs increase NO levels in ovarian and endometrial cancer cells and promote proliferation in these cells. O-ASCs and cancer cell cocultures revealed that cancer cells use O-ASC-secreted arginine and in turn secrete citrulline in the microenvironment. Interestingly, citrulline increased adipogenesis potential of the O-ASCs. Furthermore, we found that O-ASCs increased NO synthesis in cancer cells, leading to decrease in mitochondrial respiration in these cells. Our findings suggest that O-ASCs upregulate glycolysis and reduce oxidative stress in cancer cells by increasing NO levels through paracrine metabolite secretion. Significantly, we found that O-ASC-mediated chemoresistance in cancer cells can be deregulated by altering NO homeostasis. A combined approach of targeting secreted arginine through l-arginase, along with targeting microenvironment-secreted factors using l-NAME, may be a viable therapeutic approach for targeting ovarian and endometrial cancers.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
D-(+)-Glucose, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Pricing and availability is not currently available.
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Rosiglitazone
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Pricing and availability is not currently available.
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Pricing and availability is not currently available.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
2-Propanol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Pricing and availability is not currently available.