Merck
  • Selective extraction of catecholamines by packed fiber solid-phase using composite nanofibers composing of polymeric crown ether with polystyrene.
Selective extraction of catecholamines by packed fiber solid-phase using composite nanofibers composing of polymeric crown ether with polystyrene.
Biomedical chromatography : BMC (2014-06-12)
Li-Qin Chen, Yu Wang, Jian-Song Qu, Jian-Jun Deng, Xue-Jun Kang
ABSTRACT
For the first time, electrospun composite nanofibers comprising polymeric crown ether with polystyrene (PCE-PS) have been used for the selective extraction of catecholamines - dopamine (DA), norepinephrine (NE) and epinephrine (E) - prior to their analysis by high-performance liquid chromatography-electrochemical detection. Using a minicartridge packed with PCE-PS composite nanofibers, the target compounds were extracted effectively from urine samples to which diphenylborinic acid 2-aminoethyl ester was added as a complexing reagent. The extracted catecholamines could be liberated from the fiber by the addition of acetic acid. A good linearity was observed for catecholamines in the range of 2.0-200 ng mL(-1) (NE, E and DA). The detection limits of catecholamines (signal-to-noise ratio = 3) were 0.5 ng mL(-1) (NE), 0.2 ng mL(-1) (E) and 0.2 ng mL(-1) (DA), respectively. Under the optimized conditions, the absolute recoveries of the above three catecholamines were 90.6% (NE), 88.5% (E) and 94.5% (DA). The repeatability of extraction performance was from 5.4 to 9.2% (expressed as relative standard deviation). Our results indicate that the proposed method could be used for the determination of NE, E and DA in urine.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Supelco
3,4-Dihydroxyphenylacetic acid, analytical standard
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
3,4-Dihydroxybenzylamine hydrobromide, 98%
Sigma-Aldrich
Phenylboronic acid, 95%
Sigma-Aldrich
Phenylboronic acid, purum, ≥97.0% (HPLC)