Merck
  • Checkpoint kinase inhibitor AZD7762 overcomes cisplatin resistance in clear cell carcinoma of the ovary.
Checkpoint kinase inhibitor AZD7762 overcomes cisplatin resistance in clear cell carcinoma of the ovary.
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society (2013-12-24)
Hiroaki Itamochi, Mayumi Nishimura, Nao Oumi, Misaki Kato, Tetsuro Oishi, Muneaki Shimada, Shinya Sato, Jun Naniwa, Seiya Sato, Akiko Kudoh, Junzo Kigawa, Tasuku Harada
ABSTRACT
Checkpoint kinase (Chk) inhibitors are thought to increase the cytotoxic effects of DNA-damaging agents and are undergoing clinical trials. The present study was aimed to assess the potential to use the Chk1 and Chk2 inhibitor, AZD7762, with other anticancer agents in chemotherapy to treat ovarian clear cell carcinoma. Four ovarian clear cell carcinoma cell lines were used in this study. We treated the cells with AZD7762 and anticancer agents, then assessed cell viability, cell cycle distribution, apoptosis, and the expression of protein in apoptotic pathways and molecules downstream of the Chk signaling pathways. We also investigated the effects of these drug combinations on tumor growth in a nude mouse xenograft model. Synergistic effects from the combination of AZD7762 and cisplatin were observed in all 4 cell lines. However, we observed additive effects when AZD7762 was combined with paclitaxel on all cell lines tested. AZD7762 effectively suppressed the Chk signaling pathways activated by cisplatin, dramatically enhanced expression of phosphorylated H2A.X, cleaved caspase 9 and PARP, decreased the proportion of cells in the gap 0/ gap 1 phase and the synthesis-phase fraction, and increased apoptotic cells. Combinations of small interfering RNA against Chk 1 and small interfering RNA against Chk2 enhanced the cytotoxic effect of cisplatin in both RMG-I and KK cells. Finally, treating mice-bearing RMG-I with AZD7762 and cisplatin significantly suppressed growth of tumors in a xenograft model. The present study indicates that chemotherapy with AZD7762 and cisplatin should be explored as a treatment modality for women with ovarian clear cell carcinoma.
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Paclitaxel natural for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
L-Threonine, European Pharmacopoeia (EP) Reference Standard
Serine, European Pharmacopoeia (EP) Reference Standard