Merck
  • Post-column mobile phase adjustment: a strategy to eliminate the contradiction between liquid chromatography and mass spectrometry in the determination of flavonoids in rat plasma.
Post-column mobile phase adjustment: a strategy to eliminate the contradiction between liquid chromatography and mass spectrometry in the determination of flavonoids in rat plasma.
Journal of pharmaceutical and biomedical analysis (2014-03-29)
Shirui Zheng, Zhiyuan Ma, Haixia Han, Jianfeng Ye, Ruwei Wang, Sheng Cai, Hui Zhou, Lushan Yu, Su Zeng, Huidi Jiang
ABSTRACT
Flavonoids are a group of important naturally occurring polyphenolic compounds with a wide range of biological effects. In this study, a sensitive liquid chromatography tandem mass spectrometry method was developed to simultaneously determine multiple active flavonoids, including quercetin (Que), kaempferol (Kae), apigenin (Api), isorhamnetin (Iso), luteolin (Lut), and naringenin (Nar), in rat plasma. To achieve a satisfied peak shape and LC separation, formic acid with the concentration between 0.05 and 0.2%, or in some case 5%, was generally used to acidify the LC mobile phase in reported studies. Here we found that even 0.05% formic acid could lead to strong mass signal suppression, and the absence of formic acid could reverse the signal suppression but cause serious peak tailing. There is an irreconcilable contradiction between liquid chromatography (LC) and mass spectrometry (MS). In order to simultaneously satisfy LC and MS, LC mobile phase with 0.00075% formic acid and post column mobile phase adjustment with 0.0677% ammonium solution in isopropanol were applied. Compared with the conventional method with mobile phase containing 0.05% formic acid, the mass signal response of Que, Kae, Api, Iso, Lut, Nar, and Oka increased 26.2, 18.6, 13.6, 23.5, 17.5, 15.6 and 15.4 fold, respectively. In addition, the post column mobile phase addition exhibited the better peak shape for the reduction of analytes longitudinal diffusion. The method has been fully validated according to FDA guidelines within the linear range between 0.328 ng mL⁻¹ and 168 ng mL⁻¹, and successfully applied to a pilot pharmacokinetic study of rats after administering 5.43 g kg⁻¹ Pollen of Brassica campestris.
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%