Merck
  • Protein classification and distribution in osteoarthritic human synovial tissue by matrix-assisted laser desorption ionization mass spectrometry imaging.
Protein classification and distribution in osteoarthritic human synovial tissue by matrix-assisted laser desorption ionization mass spectrometry imaging.
Analytical and bioanalytical chemistry (2014-12-17)
Berta Cillero-Pastor, Gert B Eijkel, Francisco J Blanco, Ron M A Heeren
ABSTRACT
The remodeling of the synovial membrane, which normally lubricates the joints by producing synovial fluid, is one of the most characteristic events in the pathology of osteoarthritis (OA). The heterogeneity and spatial distribution of proteins in the synovial membrane are poorly studied and we hypothesized that they constitute excellent molecular disease classifiers for the accurate diagnosis of the disease. Matrix-assisted laser desorption ionization mass spectrometry imaging (MALDI-MSI) allows for the study of the localization and identification of hundreds of different molecules with high sensitivity in very thin tissue sections. In this work, we employed MALDI-MSI in combination with principal component analysis and discriminant analysis to reveal the specific profile and distribution of digested proteins in human normal and OA synovial membranes. Proteins such as hemoglobin subunit alpha 2, hemoglobin subunit beta, actin aortic smooth muscle, biglycan, and fibronectin have been directly identified from human synovial biopsies. In addition, we have determined the location of disease-specific OA markers. Some of them which are located in areas of low inflammation provide valuable information on tissue heterogeneity. Finally, we described the OA molecular protein signatures common to synovial and other articular tissues such as cartilage. For the first time, normal and OA human synovial membranes have been classified by MALDI-MSI, thus offering a new sensitive tool for the study of rheumatic pathologies.
Product Description
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Hematoxylin, certified by the BSC
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%