Merck
  • Simultaneous determination of propofol and its glucuronide in whole blood by liquid chromatography-electrospray tandem mass spectrometry and the influence of sample storage conditions on the reliability of the test results.
Simultaneous determination of propofol and its glucuronide in whole blood by liquid chromatography-electrospray tandem mass spectrometry and the influence of sample storage conditions on the reliability of the test results.
Journal of pharmaceutical and biomedical analysis (2015-03-17)
Lambert K Sørensen, Jørgen B Hasselstrøm
ABSTRACT
Propofol (2,6-diisopropylphenol) is commonly used as an anaesthetic agent but is also abused for recreational purposes. Several cases of fatalities involving self-administered propofol have been reported. For rapid quantification of propofol and propofol β-d-glucuronide (propofol G) in clinical and forensic cases, an ultra-performance liquid chromatography-tandem mass spectrometry method using pneumatically assisted electrospray ionisation has been developed. The technique has been validated on both ante-mortem and post-mortem human whole blood. The proteins in the blood samples were removed by the addition of a mixture of methanol and acetonitrile, and the extract was cleaned up by solid phase extraction. The extract was concentrated in dimethyl sulphoxide. The system was calibrated using matrix-matched calibrants combined with isotope dilution. The lower limits of quantification were 0.01 and 0.02mg/L for propofol and 0.02 and 0.04mg/L for propofol G in ante-mortem and post-mortem whole blood, respectively. The relative intra-laboratory reproducibility standard deviation was less than 10% at concentrations of 0.2mg/L or higher. The mean true extraction recovery was 85% for propofol and 81% for propofol G. The trueness of the propofol determination expressed as the relative bias of the test results was within ±6% at concentration levels of 0.01-8.5mg/L. Propofol was less stable in blood stabilised with a citrate-EDTA-fluoride mixture than in blood stabilised with an oxalate-fluoride mixture. The stability was lower at -20°C than at 5°C and -80°C. Propofol G did not show instability under the storage conditions tested.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Propofol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%