Merck
  • Novel markers of osteogenic and adipogenic differentiation of human bone marrow stromal cells identified using a quantitative proteomics approach.
Novel markers of osteogenic and adipogenic differentiation of human bone marrow stromal cells identified using a quantitative proteomics approach.
Stem cell research (2013-11-19)
Cecilia Granéli, Anna Thorfve, Ulla Ruetschi, Helena Brisby, Peter Thomsen, Anders Lindahl, Camilla Karlsson
ABSTRACT
Today, the tool that is most commonly used to evaluate the osteogenic differentiation of bone marrow stromal cells (BMSCs) in vitro is the demonstration of the expression of multiple relevant markers, such as ALP, RUNX2 and OCN. However, as yet, there is no single surface marker or panel of markers which clearly defines human BMSCs (hBMSCs) differentiating towards the osteogenic lineage. The aim of this study was therefore to examine this issue. Stable isotope labeling by amino acids in cell culture (SILAC)-based quantitative proteomics was utilized to investigate differently expressed surface markers in osteogenically differentiated and undifferentiated hBMSCs. Labeled membrane proteins were analyzed by mass spectrometry (MS) and 52 proteins with an expression ratio above 2, between osteogenically differentiated and undifferentiated cells, were identified. Subsequent validation, by flow cytometry and ELISA, of the SILAC expression ratios for a number of these proteins and investigations of the lineage specificity of three candidate markers were performed. The surface markers, CD10 and CD92, demonstrated significantly increased expression in hBMSCs differentiated towards the osteogenic and adipogenic lineages. In addition, there was a slight increase in CD10 expression during chondrogenic differentiation. Furthermore, the expression of the intracellular protein, crystalline-αB (CRYaB), was only significantly increased in osteogenically differentiated hBMSCs and not affected during differentiation towards the chondrogenic or adipogenic lineages. It has been concluded from the present results that CD10 and CD92 are potential markers of osteogenic and adipogenic differentiation and that CRYaB is a potential novel osteogenic marker specifically expressed during the osteogenic differentiation of hBMSCs in vitro.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Gram′s safranin solution, suitable for microscopy
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material