Merck
  • High IP-10 levels decrease T cell function in HIV-1-infected individuals on ART.
High IP-10 levels decrease T cell function in HIV-1-infected individuals on ART.
Journal of leukocyte biology (2014-08-27)
L A Ramirez, T A Arango, E Thompson, M Naji, P Tebas, J D Boyer
ABSTRACT
HIV-1-infected subjects, despite control of viral replication with ART, have an altered immune cytokine/chemokine milieu. Changes in systemic cytokines and chemokines can alter immune responses. IP-10, in particular, has been associated with pathogenesis in a number of conditions, and we found that IP-10 is increased in serum in subjects who are HIV-1 infected and on stable ART compared with HIV-1-uninfected individuals. In a series of in vitro studies, we found that PBMCs exposed to IP-10 showed a significant decrease in the number of cells capable of secreting IFN-γ, as well as other cytokines, when stimulated with recall antigens. Furthermore, treatment with IP-10 led to decreased antigen-specific calcium signaling and MAPK38 phosphorylation. Importantly, the cytokines, as well as proliferative responses, could be enhanced with an IP-10 Nab. Our findings suggest that IP-10-modulating drugs may potentially enhance T cell responses to vaccination and HIV-1 in HIV+ subjects on ART.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis