Merck
  • Effect of selective heparin desulfation on preservation of bone morphogenetic protein-2 bioactivity after thermal stress.
Effect of selective heparin desulfation on preservation of bone morphogenetic protein-2 bioactivity after thermal stress.
Bioconjugate chemistry (2015-01-27)
Song P Seto, Tobias Miller, Johnna S Temenoff
ABSTRACT
Bone morphogenetic protein-2 (BMP-2) plays an important role in bone and cartilage formation and is of interest in regenerative medicine. Heparin can interact electrostatically with BMP-2 and thus has been explored for controlled release and potential stabilization of this growth factor in vivo. However, in its natively sulfated state, heparin has potent anticoagulant properties that may limit its use. Desulfation reduces anticoagulant properties, but may impact heparin's ability to interact and protect BMP-2 from denaturation. The goal of this study was to characterize three selectively desulfated heparin species (N-desulfated (Hep(-N)), 6-O,N-desulfated (Hep(-N,-6O)), and completely desulfated heparin (Hep(-))) and determine if the sulfation level of heparin affected the level of BMP-2 bioactivity after heat treatment at 65 °C. BMP-2 bioactivity was evaluated using the established C2C12 cell assay. The resulting alkaline phosphatase activity data demonstrated that native heparin maintained a significant amount of BMP-2 bioactivity and the effect appeared to be heparin concentration dependent. Although all three had the same molecular charge as determined by zeta potential measurements, desulfated heparin derivatives Hep(-N) and Hep(-N,-6O) were not as effective as native heparin in maintaining BMP-2 bioactivity (only ~35% of original activity remained in both cases). These findings can be used to better select desulfated heparin species that exhibit low anticoagulant activity while extending the half-life of BMP-2 in solution and in delivery systems.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Diethyl ether
Supelco
1-Methyl-2-pyrrolidinone, analytical standard
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, suitable for HPLC, ≥99%
Sigma-Aldrich
Potassium acetate solution, BioUltra, Molecular Biology, 5 M in H2O
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Supelco
Methanol, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol