Merck
  • Genes Indicative of Zoonotic and Swine Pathogens Are Persistent in Stream Water and Sediment following a Swine Manure Spill.
Genes Indicative of Zoonotic and Swine Pathogens Are Persistent in Stream Water and Sediment following a Swine Manure Spill.
Applied and environmental microbiology (2015-03-15)
Sheridan K Haack, Joseph W Duris, Dana W Kolpin, Lisa R Fogarty, Heather E Johnson, Kristen E Gibson, Michael Focazio, Kellogg J Schwab, Laura E Hubbard, William T Foreman
ABSTRACT
Manure spills into streams are relatively frequent, but no studies have characterized stream contamination with zoonotic and veterinary pathogens, or fecal chemicals, following a spill. We tested stream water and sediment over 25 days and downstream for 7.6 km for the following: fecal indicator bacteria (FIB), the fecal indicator chemicals cholesterol and coprostanol, 20 genes for zoonotic and swine-specific bacterial pathogens by presence/absence PCR for viable cells, one swine-specific Escherichia coli toxin gene (STII gene) by quantitative PCR (qPCR), and nine human and animal viruses by qPCR or reverse transcription-qPCR. Twelve days postspill, and 4.2 km downstream, water concentrations of FIB, cholesterol, and coprostanol were 1 to 2 orders of magnitude greater than those detected before, or above, the spill, and genes indicating viable zoonotic or swine-infectious Escherichia coli were detected in water or sediment. STII gene levels increased from undetectable before or above the spill to 10(5) copies/100 ml of water 12 days postspill. Thirteen of 14 water (8/9 sediment) samples had viable STII-carrying cells postspill. Eighteen days postspill, porcine adenovirus and teschovirus were detected 5.6 km downstream. FIB concentrations (per gram [wet weight]) in sediment were greater than in water, and sediment was a continuous reservoir of genes and chemicals postspill. Constituent concentrations were much lower, and detections less frequent, in a runoff event (200 days postspill) following manure application, although the swine-associated STII and stx2e genes were detected. Manure spills are an underappreciated pathway for livestock-derived contaminants to enter streams, with persistent environmental outcomes and the potential for human and veterinary health consequences.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)