Merck
  • Vaccines against human HER2 prevent mammary carcinoma in mice transgenic for human HER2.
Vaccines against human HER2 prevent mammary carcinoma in mice transgenic for human HER2.
Breast cancer research : BCR (2014-01-24)
Carla De Giovanni, Giordano Nicoletti, Elena Quaglino, Lorena Landuzzi, Arianna Palladini, Marianna Lucia Ianzano, Massimiliano Dall'Ora, Valentina Grosso, Dario Ranieri, Roberta Laranga, Stefania Croci, Augusto Amici, Manuel L Penichet, Manuela Iezzi, Federica Cavallo, Patrizia Nanni, Pier-Luigi Lollini
ABSTRACT
The availability of mice transgenic for the human HER2 gene (huHER2) and prone to the development of HER2-driven mammary carcinogenesis (referred to as FVB-huHER2 mice) prompted us to study active immunopreventive strategies targeting the human HER2 molecule in a tolerant host. FVB-huHER2 mice were vaccinated with either IL-12-adjuvanted human HER2-positive cancer cells or DNA vaccine carrying chimeric human-rat HER2 sequences. Onset and number of mammary tumors were recorded to evaluate vaccine potency. Mice sera were collected and passively transferred to xenograft-bearing mice to assess their antitumor efficacy. Both cell and DNA vaccines significantly delayed tumor onset, leading to about 65% tumor-free mice at 70 weeks, whereas mock-vaccinated FVB-huHER2 controls developed mammary tumors at a median age of 45 weeks. In the DNA vaccinated group, 65% of mice were still tumor-free at about 90 weeks of age. The number of mammary tumors per mouse was also significantly reduced in vaccinated mice. Vaccines broke the immunological tolerance to the huHER2 transgene, inducing both humoral and cytokine responses. The DNA vaccine mainly induced a high and sustained level of anti-huHER2 antibodies, the cell vaccine also elicited interferon (IFN)-γ production. Sera of DNA-vaccinated mice transferred to xenograft-carrying mice significantly inhibited the growth of human HER2-positive cancer cells. Anti-huHER2 antibodies elicited in the tolerant host exert antitumor activity.
Product Description
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
USP
Methyl salicylate, United States Pharmacopeia (USP) Reference Standard
Supelco
Methyl salicylate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Supelco
Methyl salicylate, analytical standard
Sigma-Aldrich
Methyl salicylate, ≥98%, FCC, FG
Sigma-Aldrich
Methyl salicylate, ReagentPlus®, ≥99% (GC)
Sigma-Aldrich
Methyl salicylate, tested according to Ph. Eur.
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC