Merck
Label-free monitoring of μ-opioid receptor-mediated signaling.
Molecular pharmacology (2014-05-31)
Philippe Bourassa, Hanieh Bagheri Tudashki, Graciela Pineyro, Michel Grandbois, Louis Gendron
ABSTRACT
In this study, we used a combination of traditional signaling investigation approaches, bioluminescence resonance energy transfer (BRET) biosensors, and the label-free approach surface plasmon resonance (SPR) spectroscopy to monitor the signaling cascades of the μ-opioid receptor (MOP). In human embryonic kidney cells stably expressing a Flag-tagged version of human MOP, we compared the signals triggered by the noninternalizing and internalizing MOP agonists morphine and DAMGO (Tyr-D-Ala-Gly-N-methyl-Phe-Gly-ol), respectively. We studied three major and well described components of MOP signaling: receptor internalization, G protein coupling, and activation of extracellular signal-regulated kinase ERK1/ERK2. Our results show that morphine and DAMGO display different profiles of receptor internalization and a similar ability to trigger the phosphorylation of ERK1/ERK2. Our SPR analyses revealed that morphine and DAMGO evoke similar SPR signatures and that Gαi, cAMP-dependent pathways, and ERK1/ERK2 have key roles in morphine- and DAMGO-mediated signaling. Most interestingly, we found that the so-called MOP neutral antagonists CTOP (D-Phe-Cys-Tyr-D-Trp-Orn-Thr-Pen-Thr-NH(2)), naloxone, and naltrexone behave like partial agonists. Even more intriguing, BRET experiments indicate that CTAP (D-Phe-Cys-Tyr-D-Trp-Arg-Thr-Pen-Thr-NH(2)) induces similar conformational changes as naltrexone at the Gαi-βγ interface, whereas it appears as an inverse agonist based on its SPR response thus indicating distinct signaling mechanisms for the two ligands. Taken together, our results support the usefulness of label-free methods such as SPR to study whole-cell responses and signaling cascades triggered by G protein-coupled receptors and complement the conventional approaches by revealing cellular responses that would have been otherwise undetectable.
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Staurosporine, VETRANAL®, analytical standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®