Merck
  • In vitro chondrocyte behavior on porous biodegradable poly(e-caprolactone)/polyglycolic acid scaffolds for articular chondrocyte adhesion and proliferation.
In vitro chondrocyte behavior on porous biodegradable poly(e-caprolactone)/polyglycolic acid scaffolds for articular chondrocyte adhesion and proliferation.
Journal of biomaterials science. Polymer edition (2015-02-12)
John B Jonnalagadda, Iris V Rivero, Janet S Dertien
ABSTRACT
In this study, poly(e-caprolactone)/polyglycolic acid (PCL/PGA) scaffolds for repairing articular cartilage were fabricated via solid-state cryomilling along with compression molding and porogen leaching. Four distinct scaffolds were fabricated using this approach by four independent cryomilling times. These scaffolds were assessed for their suitability to promote articular cartilage regeneration with in vitro chondrocyte cell culture studies. The scaffolds were characterized for pore size, porosity, swelling ratio, compressive, and thermal properties. Cryomilling time proved to significantly affect the physical, mechanical, and morphological properties of the scaffolds. In vitro bovine chondrocyte culture was performed dynamically for 1, 7, 14, 28, and 35 days. Chondrocyte viability and adhesion were tested using MTT assay and scanning electron microscopy micrographs. Glycosaminoglycan (GAG) and DNA assays were performed to investigate the extracellular matrix (ECM) formation and cell proliferation, respectively. PCL/PGA scaffolds demonstrated high porosity for all scaffold types. Morphological analysis and poly(ethylene oxide) continuity demonstrated the existence of a co-continuous network of interconnected pores with pore sizes appropriate for tissue engineering and chondrocyte ingrowth. While mean pore size decreased, water uptake and compressive properties increased with increasing cryomilling times. Compressive modulus of 12, 30, and 60 min scaffolds matched the compressive modulus of human articular cartilage. Viable cells increased besides increase in cell proliferation and ECM formation with progress in culture period. Chondrocytes exhibited spherical morphology on all scaffold types. The pore size of the scaffold affected chondrocyte adhesion, proliferation, and GAG secretion. The results indicated that the 12 min scaffolds delivered promising results for applications in articular cartilage repair.
Product Description
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
SAFC
L-Proline
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture