Merck
  • Squamous tissue lymphocytes in the esophagus of controls and patients with reflux esophagitis and Barrett's esophagus are characterized by a non-inflammatory phenotype.
Squamous tissue lymphocytes in the esophagus of controls and patients with reflux esophagitis and Barrett's esophagus are characterized by a non-inflammatory phenotype.
PloS one (2014-08-30)
Alexandra Lind, Leo Koenderman, Johannes G Kusters, Peter D Siersema
ABSTRACT
Reflux esophagitis (RE) is characterized by inflammation of the squamous epithelium (SQ) of the esophagus and may progress to Barrett's esophagus (BE) characterized by intestinal metaplasia. The role of inflammation in this transition has been postulated but lacks experimental evidence. Here, the inflammatory responses in the esophagus of these patients were investigated. Fifty-one esophageal biopsies from with patients BE (n = 19), RE (n = 8) and controls (n = 23) were analyzed. T-cells were analyzed before and after ex vivo expansion (14 days) by multicolor flow cytometric analysis. The following markers were studied: CD3, CD4, CD8 (T-cell markers), Granzyme B (marker of cytotoxicity), CD103 (αE/epithelial integrin) and NKg2a (inhibitory receptor on T-cells and NK-cells). Analysis of ex vivo cultures from normal looking SQ from controls, RE patients, and BE patients revealed no significant differences in the number and phenotypes of T-cells. In contrast, tissue from RE was different to normal SQ in four aspects: 1) higher percentages of CD3+ CD4+-cells (72±7% vs 48±6%, p = 0.01) and 2) CD8+ GranzymeB+-cells (53±11% vs 26±4%, p<0.05), while 3) lower percentages of CD4+ CD103+-cells (45±19% vs 80±3%, p = 0.02) and 4) CD8+ NKg2a+-cells (31±12% vs 44±5%). Despite the fact that both tissues are exposed to the same reflux associated inflammatory triggers, the immune response observed in RE is clearly distinct from that in SQ of BE. The differences in immune responses in BE tissue might contribute to its susceptibility for transformation into intestinal metaplasia.
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland