Merck
  • Intranasal chitosan-DNA vaccines that protect across influenza virus subtypes.
Intranasal chitosan-DNA vaccines that protect across influenza virus subtypes.
International journal of pharmaceutics (2014-07-08)
Chompoonuch Sawaengsak, Yasuko Mori, Koichi Yamanishi, Potjanee Srimanote, Wanpen Chaicumpa, Ampol Mitrevej, Nuttanan Sinchaipanid
ABSTRACT
An egg-free and broadly effective influenza vaccine that can be produced rapidly, adequately and cost-effectively is needed. In this study, chitosan-associated DNAs prepared at various nitrogen/phosphate charge (N/P) ratios were studied for their physicochemical properties, stability, cytotoxicity, and protein expression ability. The chitosan-DNA complexes (chitoplexes) of the N/P ratio 2 had the required characteristics including optimal size range, positive surface charge, high DNA association efficiency, tolerability to DNase digestion and mammalian cell viability compatibility. The N/P ratio 2-chitoplexes revealed the highest green fluorescent protein and luciferase expressions in the transfected mammalian cell cultures and in the mouse lungs. Mice immunized intranasally with the N/P ratio 2-chitoplex vaccines carrying DNAs coding for conserved proteins of influenza virus, i.e., ion channel protein (M2) and/or nucleoprotein (NP), had both mucosal and systemic humoral as well as cell mediated immune responses to the in vivo expressed antigens which conferred protection in mice against the lethal challenges not only with the homologous influenza virus subtype (H1N1), but also the heterologous subtype (H3N2). The chitoplexes should be considered as influenza vaccine candidates especially during the period of high vaccine demand. They are suitable for developing areas of the world where conventional vaccine production capacity is lacking.
Product Description
Millipore
Bifido Selective Supplement B, suitable for microbiology
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%