Merck
  • Nox2-dependent signaling between macrophages and sensory neurons contributes to neuropathic pain hypersensitivity.
Nox2-dependent signaling between macrophages and sensory neurons contributes to neuropathic pain hypersensitivity.
Pain (2014-08-21)
Wiebke Kallenborn-Gerhardt, Stephan W Hohmann, Katharina M J Syhr, Katrin Schröder, Marco Sisignano, Andreas Weigert, Jana E Lorenz, Ruirui Lu, Bernhard Brüne, Ralf P Brandes, Gerd Geisslinger, Achim Schmidtko
ABSTRACT
Emerging lines of evidence indicate that production of reactive oxygen species (ROS) at distinct sites of the nociceptive system contributes to the processing of neuropathic pain. However, the mechanisms underlying ROS production during neuropathic pain processing are not fully understood. We here detected the ROS-generating nicotinamide adenine dinucleotide phosphate oxidase isoform Nox2 in macrophages of dorsal root ganglia (DRG) in mice. In response to peripheral nerve injury, Nox2-positive macrophages were recruited to DRG, and ROS production was increased in a Nox2-dependent manner. Nox2-deficient mice displayed reduced neuropathic pain behavior after peripheral nerve injury, whereas their immediate responses to noxious stimuli were normal. Moreover, injury-induced upregulation of tumor necrosis factor α was absent, and activating transcription factor 3 induction was reduced in DRG of Nox2-deficient mice, suggesting an attenuated macrophage-neuron signaling. These data suggest that Nox2-dependent ROS production in macrophages recruited to DRG contributes to neuropathic pain hypersensitivity, underlining the observation that Nox-derived ROS exert specific functions during the processing of pain.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland