Merck
  • Inhibition of human efflux transporter ABCC2 (MRP2) by self-emulsifying drug delivery system: influences of concentration and combination of excipients.
Inhibition of human efflux transporter ABCC2 (MRP2) by self-emulsifying drug delivery system: influences of concentration and combination of excipients.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques (2015-01-13)
Liang Li, Tao Yi, Christopher Wai-Kei Lam
ABSTRACT
This study investigated influences of concentration and combination of excipients, commonly used in self-emulsifying drug delivery systems (SEDDS), on inhibition of human efflux transporter ABCC2 (MRP2). Ten commonly used excipients of SEDDS with inhibitory effect on MRP2 including Cremophor® EL, Cremophor® RH, Pluronic® F127, Maisine® 35-1, β-cyclodextrin, Labrasol®, Pluronic® F68, PEG 2000, PEG 400 and Transcutol® were studied with the Caco-2 cell model. Six excipients with inhibitory effect including Cremophor® EL, Cremophor® RH, Pluronic® F127, PEG 2000, PEG 400 and Transcutol® were further analyzed using the MRP2 vesicle assay and ATPase activity assay. Ultra-performance liquid-chromatography tandem mass spectrometry was used to measure scutellarin as the MRP2 substrate. In studying concentration-dependent effects, five excipients including Cremophor® EL, Cremophor® RH, Pluronic® F127, Maisine® 35-1 and β-cyclodextrin showed concentration-dependent decrease in efflux ratio of scutellarin. The other five excipients did not show such phenomenon, and their inhibitory effects were restricted to be above to certain critical or minimum concentrations. In studying combined effects, PEG 2000 and Pluronic® F127 both showed combined effect with Cremophor® EL on inhibiting MRP2. However, some combinations of excipients such as PEG 400 and Transcutol® with Cremophor® EL increased the scutellarin efflux ratio and decreased the transport of scutellarin and ATPase activity, compared to Cremophor® EL alone. The above results suggest that appropriate choice of excipients according to their concentration-dependent and combined effects on MRP2 inhibition can facilitate formulation of SEDDS for improving the bioavailability of drugs that are MRP2 substrates.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetic acid, analytical standard
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Piperazine, ReagentPlus®, 99%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethanesulfonic acid, 95%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%