Merck
  • Vaginal expression of efflux transporters and the potential impact on the disposition of microbicides in vitro and in rabbits.
Vaginal expression of efflux transporters and the potential impact on the disposition of microbicides in vitro and in rabbits.
Molecular pharmaceutics (2014-10-21)
Carolien Grammen, Myriam Baes, Steven Haenen, Jasper Verguts, Koen Augustyns, Thomas Zydowsky, Paolo La Colla, Patrick Augustijns, Joachim Brouwers
ABSTRACT
In order to reach sufficiently high tissue concentrations and thus be effective, vaginally applied anti-HIV microbicides that are active at the level of the immune cells must permeate across the cervicovaginal mucosal layer. Cellular efflux transporters, such as Pgp, BCRP, and MRP-2, have been demonstrated to greatly affect drug disposition at different sites in the body including the intestine and the blood-brain barrier; their possible role on drug uptake from the female genital tract, however, has not been elucidated yet. In the present study, the protein expression of Pgp, BCRP, and MRP-2 in endocervical and vaginal tissue of premenopausal women was confirmed by Western blot analysis. To enable the assessment of transporter effects in vitro, the identification of an appropriate cervicovaginal cell line was pursued. The cervical SiHa cell line was observed to express mRNA of the 3 studied transporters, but only MRP-2 was found to be active. Consequently, the established Caco-2 cell line was utilized as an alternative in which the interaction of 10 microbicide candidates with the efflux transporters was studied. Darunavir, saquinavir, and maraviroc were identified as Pgp and MRP-2 substrates. The impact of Pgp on in vivo drug disposition was further examined for the model Pgp substrate talinolol in rabbits. Its vaginal uptake was significantly reduced by Pgp-mediated efflux when formulated in a neutral but not in an acidic gel. Our findings indicate the expression of a functional Pgp transporter in the vaginal mucosa that may severely reduce the vaginal uptake of Pgp substrates, including certain microbicide candidates, especially in women with an increased vaginal pH.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%