Merck
  • Survey regarding the occurrence of selected organic micropollutants in the groundwaters of overseas departments.
Survey regarding the occurrence of selected organic micropollutants in the groundwaters of overseas departments.
Environmental science and pollution research international (2014-03-07)
Emmanuelle Vulliet, Mikaël Tournier, Antoine Vauchez, Laure Wiest, Robert Baudot, Florent Lafay, Agneta Kiss, Cécile Cren-Olivé
ABSTRACT
To collect a complete dataset regarding the occurrence of organic substances in groundwater, this study presents the examination of 66 organic contaminants in the groundwater of overseas departments, including pesticides, pharmaceutical compounds, hormones and some industrial substances. The selective and sensitive analytical methods are described. These techniques begin with solid-phase extraction (SPE) followed by analysis using liquid chromatography-tandem mass spectrometry (LC-MS/MS) and gas chromatography-mass spectrometry (GC-ToF-MS). The paper summarises the analytical results from 40 sampling points collected during two campaigns in Guadeloupe, Martinique, Reunion, Mayotte and Guiana, representing 80 samples. Of the 66 target substances, 36 were determined at least once. Among the most frequently detected are bisphenol A (frequency, 96%; max., 7,400 ng/L), caffeine (frequency, 91%; max., 1,240 ng/L), pentachlorophenol (frequency, 55%; max., 418 ng/L), and carbamazepine (frequency, 56%; max., 22 ng/L). The results do not put in evidence that the origin of the sample or climatic characteristics of these regions influence the dilution and release of micropollutants.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis