Merck
  • Suitability of porcine chondrocyte micromass culture to model osteoarthritis in vitro.
Suitability of porcine chondrocyte micromass culture to model osteoarthritis in vitro.
Molecular pharmaceutics (2014-03-19)
Niels Schlichting, Tilo Dehne, Karsten Mans, Michaela Endres, Bruno Stuhlmüller, Michael Sittinger, Christian Kaps, Jochen Ringe
ABSTRACT
In vitro tissue models are useful tools for the development of novel therapy strategies in cartilage repair and care. The limited availability of human primary tissue and high costs of animal models hamper preclinical tests of innovative substances and techniques. In this study we tested the potential of porcine chondrocyte micromass cultures to mimic human articular cartilage and essential aspects of osteoarthritis (OA) in vitro. Primary chondrocytes were enzymatically isolated from porcine femoral condyles and were maintained in 96-multiwell format to establish micromass cultures in a high-throughput scale. Recombinant porcine tumor necrosis factor alpha (TNF-α) was used to induce OA-like changes documented on histological (Safranin O, collagen type II staining), biochemical (hydroxyproline assay, dimethylmethylene blue method), and gene expression level (Affymetrix porcine microarray, real time PCR) and were compared with published data from human articular cartilage and human micromass cultures. After 14 days in micromass culture, porcine primary chondrocytes produced ECM rich in proteoglycans and collagens. On gene expression level, significant correlations of detected genes with porcine cartilage (r = 0.90), human cartilage (r = 0.71), and human micromass culture (r = 0.75) were observed including 34 cartilage markers such as COL2A1, COMP, and aggrecan. TNF-α stimulation led to significant proteoglycan (-75%) and collagen depletion (-50%). Comparative expression pattern analysis revealed the involvement of catabolic enzymes (MMP1, -2, -13, ADAM10), chemokines (IL8, CCL2, CXCL2, CXCL12, CCXL14), and genes associated with cell death (TNFSF10, PMAIPI, AHR) and skeletal development (GPNMB, FRZB) including transcription factors (WIF1, DLX5, TWIST1) and growth factors (IGFBP1, -3, TGFB1) consistent with published data from human OA cartilage. Expression of genes related to cartilage ECM formation (COL2A1, COL9A1, COMP, aggrecan) as well as hypertrophic bone formation (COL1A1, COL10A1) was predominantly found decreased. These findings indicating significant parallels between human articular cartilage and the presented porcine micromass model and vice versa confirm the applicability of known cartilage marker and their characteristics in the porcine micromass model. TNF-α treatment enabled the initiation of typical OA reaction patterns in terms of extensive ECM loss, cell death, formation of an inflammatory environment through the induction of genes coding for chemokines and enzymes, and the modulation of genes involved in skeletal development such as growth factors, transcription factors, and cartilage ECM-forming genes. In conclusion, the porcine micromass model represents an alternative tissue platform for the evaluation of innovative substances and techniques for the treatment of OA.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
L-Ascorbic acid, analytical standard
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade