Merck
  • Room-temperature ionic liquids enhanced green synthesis of β-glycosyl 1-ester.
Room-temperature ionic liquids enhanced green synthesis of β-glycosyl 1-ester.
Carbohydrate research (2015-02-24)
Yanli Cui, Minghan Xu, Weirong Yao, Jianwei Mao
ABSTRACT
We herein report an efficient synthesis of β-glycosyl 1-ester in room-temperature ionic liquids (RTILs) promoted via silver salt and quaternary ammonium salt (PTC) with good or excellent yields. All products were isolated exclusively as the β-anomers. Four different RTILs, eight metal salts and four quaternary ammonium salts were screened in the glycosylation reaction. The synergistic effect of C6mim·OTf, Ag2O and tetrabutylammonium iodine gave the best results. Their promotion to the system was integral. Thorough study provided insight into the catalytic activity of ionic liquid structure, metal salts and quaternary ammonium salt to these reactions. It is worth mentioning that the yield of aliphatic compound 2,3,4,6-tetra-O-acetyl-β-D-galactopyranosyl butyrate (3l) was highly improved when using C6mim·OTf as solvent compared with the normal volatile solvents under the same catalysts. This green approach has been proved to be practical and compatible with a wide range from aliphatic to aromatic substrates.
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered