Merck
  • Prednisolone-loaded nanocapsules as ocular drug delivery system: development, in vitro drug release and eye toxicity.
Prednisolone-loaded nanocapsules as ocular drug delivery system: development, in vitro drug release and eye toxicity.
Journal of microencapsulation (2014-04-05)
Tatiele Katzer, Paula Chaves, Andressa Bernardi, Adriana Pohlmann, Silvia S Guterres, Ruy Carlos Ruver Beck
ABSTRACT
To develop non-toxic aqueous ocular drug delivery systems containing prednisolone by means of its nanoencapsulation. Nanocapsules were prepared by interfacial deposition of preformed polymer [poly(ε-caprolactone) or Eudragit® RS100]. Particle size distribution was determined by laser diffractometry, photon correlation spectroscopy and nanoparticle tracking analysis. Ocular irritation and cytotoxicity were evaluated in vitro on the chorioallantoic membrane (CAM) and rabbit corneal epithelial cell line, respectively. Nanocapsules showed mean particle sizes between 100 and 300 nm and prednisolone encapsulation efficiency of around 50%. Controlled release of prednisolone occurred for 5 h for both formulations according to the biexponential model. Both formulations were found to be non-irritant in the CAM test and non-cytotoxic toward rabbit corneal epithelial cells. Encapsulation of prednisolone in nanocapsules was reported for the first time, being suitable for producing eye drops for the treatment of ocular inflammatory and no eye toxicity was indicated.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
SAFC
L-Glutamine
Sigma-Aldrich
Sorbitan monostearate, meets FCC analytical specifications
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets