Merck
The genomic landscape of pediatric Ewing sarcoma.
Cancer discovery (2014-09-05)
Brian D Crompton, Chip Stewart, Amaro Taylor-Weiner, Gabriela Alexe, Kyle C Kurek, Monica L Calicchio, Adam Kiezun, Scott L Carter, Sachet A Shukla, Swapnil S Mehta, Aaron R Thorner, Carmen de Torres, Cinzia Lavarino, Mariona Suñol, Aaron McKenna, Andrey Sivachenko, Kristian Cibulskis, Michael S Lawrence, Petar Stojanov, Mara Rosenberg, Lauren Ambrogio, Daniel Auclair, Sara Seepo, Brendan Blumenstiel, Matthew DeFelice, Ivan Imaz-Rosshandler, Angela Schwarz-Cruz Y Celis, Miguel N Rivera, Carlos Rodriguez-Galindo, Mark D Fleming, Todd R Golub, Gad Getz, Jaume Mora, Kimberly Stegmaier
ABSTRACT
Pediatric Ewing sarcoma is characterized by the expression of chimeric fusions of EWS and ETS family transcription factors, representing a paradigm for studying cancers driven by transcription factor rearrangements. In this study, we describe the somatic landscape of pediatric Ewing sarcoma. These tumors are among the most genetically normal cancers characterized to date, with only EWS-ETS rearrangements identified in the majority of tumors. STAG2 loss, however, is present in more than 15% of Ewing sarcoma tumors; occurs by point mutation, rearrangement, and likely nongenetic mechanisms; and is associated with disease dissemination. Perhaps the most striking finding is the paucity of mutations in immediately targetable signal transduction pathways, highlighting the need for new therapeutic approaches to target EWS-ETS fusions in this disease. We performed next-generation sequencing of Ewing sarcoma, a pediatric cancer involving bone, characterized by expression of EWS-ETS fusions. We found remarkably few mutations. However, we discovered that loss of STAG2 expression occurs in 15% of tumors and is associated with metastatic disease, suggesting a potential genetic vulnerability in Ewing sarcoma.
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland