Merck
  • Evaluation of 4-substituted styrenes as functional monomers for the synthesis of theophylline-specific molecularly imprinted polymers.
Evaluation of 4-substituted styrenes as functional monomers for the synthesis of theophylline-specific molecularly imprinted polymers.
Organic & biomolecular chemistry (2014-06-17)
Hazit Zayas, Clovia I Holdsworth, Michael C Bowyer, Adam McCluskey
ABSTRACT
Six novel functional monomers: 4-(4-vinylphenyl)pyridine (M1), 4′-vinylbiphenyl-4-ol (M2), N,N-dimethyl-4′-vinylbiphenyl-3-amine (M3), (4′-vinylbiphenyl-4-yl)methanol (M4), 4′-vinylbiphenyl-4-carboxylic acid (M5) and 4-hydroxy-5-methyl-4′-vinylbiphenyl-3-carboxylic acid (M6), were examined for their ability to imprint theophylline (1). Using a molecular modelling-NMR titration approach, M2 and M6 were predicted to give rise to the most specific molecularly imprinted polymers (MIPs). Rebinding analysis suggests that no imprinting effect resulted from the polymerisation of monomers M1, M5 and M6, but modest to good levels of imprinting were evident from monomers M2, M3 and M4 with IF values ranging from 1.1 (MIPM3, 20 mg) to 45 (MIPM2, 10 mg). The selective recognition of 1 varied as a function of polymer mass used. At low polymer loadings MIPM2 gave the very high IF of 45, reducing to IF = 4.1–2.3 at 20–40 mg polymer loading. With monomer M2, microwave synthesised MIP (MW-MIPM2) was examined. The MW-MIPM2 displayed lower specific rebinding than its conventionally produced counterpart (MIPM2) with IF values ranging from 1.6–2.3 (cf., MIPM2 IF 2.3–45), but significantly higher levels of rebinding with 25–52% of 1 rebound from a 0.080 mM CH3CN solution of 1 (cf., MIPM2 5–25%). MW-MIPM2 displayed a lower BET surface area than MIPM2 (185 m(2) g(−1)vs. 240 m(2) g(−1)), and lower surface (zeta) potential (−13.1 ± 8.22 mV vs. −31.4 ± 4.84 mV). Freundlich isotherm analysis revealed that MW-MIPM2 possessed higher affinity binding sites for 1 than MIPM2 with Kd values of 1.38 and 2.31 respectively. In addition, MW-MIPM2 also exhibits a higher number of binding sites (NT) compared to MW-NIPM2 (0.72 and 0.41 mg g(−1), respectively). In specificity studies using caffeine (2), MIPM2 displayed a two-fold preference for rebinding of 1 and MW-MIPM2 a five-fold preference for 1 over 2. The quantity of 2 bound in both cases was consistent with non-specific binding events. In competitive rebinding experiments, increased discrimination in favour of 1 over 2 was observed.
Product Description
USP
Theophylline, United States Pharmacopeia (USP) Reference Standard
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), purum, ≥98.0% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Theophylline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®