Merck
  • Regional lung tissue changes with mechanical ventilation and fluid load.
Regional lung tissue changes with mechanical ventilation and fluid load.
Experimental lung research (2015-04-07)
Cristiana Marcozzi, Andrea Moriondo, Eleonora Solari, Marcella Reguzzoni, Paolo Severgnini, Marina Protasoni, Alberto Passi, Paolo Pelosi, Daniela Negrini
ABSTRACT
To investigate the regional gravity-dependent impact of mechanical ventilation and fluid overload on lung extracellular matrix (ECM) in healthy lungs. The glycosaminoglycans (GAGs) composition of the ventral and dorsal lung parenchyma was determined in anesthetized supine healthy rats mechanically ventilated for 4 hours in air: (a) at low (∼7.5 mL/kg) or high (∼ 23 mL /kg) tidal volume (V(T)) and 0 cmH2O positive end-expiratory pressure (PEEP); (b) at low or high V(T) at 5 cmH2O PEEP and (c) with or without 7 mL /(kg·h) intravenous saline infusion. Mechanical ventilation degraded lung ECM, with alveolar septa thinning and structural GAGs disorganization. Low V(T) ventilation was associated with significant tissue structure changes in both ventral and dorsal lung regions, while high VT mainly affected the dependent ones. PEEP decreased ECM injury mainly in the ventral lung regions, although it did not prevent matrix fragmentation and washout at high V(T). Intravascular fluid load increased lung damage prevalently in the ventral lung regions. Mechanical ventilation and fluid load may cause additive injuries in healthy lungs, mainly in ventral regions.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Pancuronium bromide
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Pancuronium bromide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Pancuronium bromide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous