Merck
  • Anthocyanins from fermented berry beverages inhibit inflammation-related adiposity response in vitro.
Anthocyanins from fermented berry beverages inhibit inflammation-related adiposity response in vitro.
Journal of medicinal food (2014-08-01)
Diego F Garcia-Diaz, Michelle H Johnson, Elvira G de Mejia
ABSTRACT
Increased adiposity has been associated with macrophage infiltration into the adipose tissue which, in turn, leads to obesity comorbidities, including type 2 diabetes. The objective of this study was to evaluate the effect of anthocyanin (ANC)-enriched fractions from blackberry-blueberry beverages on inflammation and adipogenesis in an in vitro model of inflammation mimicking the pathologic interaction between adipocytes and macrophages. Blend ANCs inhibited secretion of nitric oxide (17.5%), tumor necrosis factor-alpha (TNF-α) (89.4%), and phosphorylated-p65 nuclear factor kappa-B (52.1%) in lipopolysaccharide (LPS)-induced RAW264.7 macrophages after 24 h. Blends reduced intracellular fat accumulation (28.2%) when applied during 3T3-L1 adipocyte differentiation and inhibited isoproterenol-induced lipolysis (18.6%) of mature 3T3-L1 cells. In addition, blend ANCs restored adiponectin-blunted gene expression induced by the TNF-α treatment (18.2%) and reduced the glycerol release (15.9%) induced by LPS-induced macrophage-conditioned media (CM) in adipocytes. Furthermore, blends slightly restored the insulin-induced glucose uptake of adipocytes, blunted by the CM treatment. In conclusion, ANCs from blueberry and blackberry dealcoholized fermented beverages are potential inhibitors of inflammation-related adiposity response and sensitizers of insulin signaling in adipocytes.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Glutamine
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture